Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000083.2 | NP_000074.2 | 16 - 17 | 2020 | 150 |
NR_046453.1 | 15 - 16 | 1873 | 150 | ||
XM_011515782.2 | XP_011514084.1 | 6 - 7 | 768 | 150 | |
XM_017011739.1 | XP_016867228.1 | 13 - 14 | 1745 | 150 | |
XM_017011740.1 | XP_016867229.1 | 13 - 14 | 1721 | 150 | |
GenBank mRNA | BC112156.1 | - | 16 - 17 | 1985 | 150 |
BC113495.1 | - | 16 - 17 | 1985 | 150 | |
BC143796.1 | - | 15 - 16 | 1838 | 150 | |
M97820.1 | - | 13 - 14 | 1424 | 150 | |
Z25884.1 | - | 16 - 17 | 2020 | 150 |