Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000083.2 | NP_000074.2 | 8 - 9 | 1062 | 99 |
NR_046453.1 | 8 - 9 | 1065 | 99 | ||
XM_017011739.1 | XP_016867228.1 | 5 - 6 | 763 | 99 | |
XM_017011740.1 | XP_016867229.1 | 5 - 6 | 763 | 99 | |
GenBank mRNA | BC112156.1 | - | 8 - 9 | 1027 | 99 |
BC113495.1 | - | 8 - 9 | 1027 | 99 | |
BC143796.1 | - | 8 - 9 | 1030 | 99 | |
M97820.1 | - | 5 - 6 | 466 | 99 | |
Z25884.1 | - | 8 - 9 | 1062 | 99 |