Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000547.5 | NP_000538.3 | 7 - 8 | 906 | 90 |
NM_001206744.1 | NP_001193673.1 | 7 - 8 | 901 | 90 | |
NM_001206745.1 | NP_001193674.1 | 7 - 8 | 901 | 90 | |
NM_175719.3 | NP_783650.1 | 7 - 8 | 906 | 90 | |
NM_175721.3 | NP_783652.1 | 6 - 7 | 816 | 90 | |
XM_011510379.2 | XP_011508681.1 | 6 - 7 | 816 | 90 | |
XM_011510380.2 | XP_011508682.1 | 6 - 7 | 816 | 90 | |
XM_011510381.2 | XP_011508683.1 | 6 - 7 | 816 | 90 | |
GenBank mRNA | AB208960.1 | - | 4 - 5 | 602 | 90 |
BC095448.1 | - | 7 - 8 | 899 | 90 | |
J02969.1 | - | 7 - 8 | 887 | 90 | |
J02970.1 | - | 7 - 8 | 887 | 90 | |
M17755.2 | - | 7 - 8 | 899 | 90 | |
X17358.1 | - | 7 - 8 | 880 | 90 | |
Y00406.1 | - | 7 - 8 | 855 | 90 |