Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127660.1 | NP_001121132.1 | 5 - 6 | 906 | 67 |
NM_014874.3 | NP_055689.1 | 6 - 7 | 1051 | 67 | |
XM_005263543.3 | XP_005263600.1 | 7 - 8 | 1064 | 67 | |
XM_005263545.3 | XP_005263602.1 | 6 - 7 | 914 | 67 | |
XM_005263547.3 | XP_005263604.1 | 6 - 7 | 946 | 67 | |
XM_005263548.3 | XP_005263605.1 | 7 - 8 | 1081 | 67 | |
GenBank mRNA | AF036536.2 | - | 6 - 7 | 920 | 67 |
AK289828.1 | - | 5 - 6 | 851 | 67 | |
AY028429.1 | - | 5 - 6 | 624 | 67 | |
BC017061.1 | - | 6 - 7 | 781 | 67 | |
D86987.1 | - | 6 - 7 | 920 | 67 |