Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127660.1 | NP_001121132.1 | 6 - 7 | 1022 | 67 |
NM_014874.3 | NP_055689.1 | 7 - 8 | 1167 | 67 | |
XM_005263543.3 | XP_005263600.1 | 8 - 9 | 1180 | 67 | |
XM_005263545.3 | XP_005263602.1 | 7 - 8 | 1030 | 67 | |
XM_005263547.3 | XP_005263604.1 | 7 - 8 | 1062 | 67 | |
XM_005263548.3 | XP_005263605.1 | 8 - 9 | 1197 | 67 | |
GenBank mRNA | AF036536.2 | - | 7 - 8 | 1036 | 67 |
AK289828.1 | - | 6 - 7 | 967 | 67 | |
AK295890.1 | - | 5 - 6 | 494 | 67 | |
AY028429.1 | - | 6 - 7 | 740 | 67 | |
BC017061.1 | - | 7 - 8 | 897 | 67 | |
D86987.1 | - | 7 - 8 | 1036 | 67 |