Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080790.1 | NP_001074259.1 | 2 - 3 | 150 | 62 |
XM_005278192.3 | XP_005278249.1 | 2 - 3 | 165 | 62 | |
GenBank mRNA | DB015621.1 | - | 2 - 3 | 150 | 62 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005365.4 | NP_005356.1 | 2 - 3 | 143 | 62 |
XM_005262334.3 | XP_005262391.1 | 2 - 3 | 165 | 62 | |
GenBank mRNA | AK292272.1 | - | 2 - 3 | 150 | 62 |
AK313321.1 | - | 2 - 3 | 150 | 62 | |
BC002351.2 | - | 2 - 3 | 143 | 62 | |
BC098100.1 | - | 2 - 3 | 104 | 62 |