Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000725.3 | NP_000716.2 | 4 - 5 | 866 | 80 |
NM_001206915.1 | NP_001193844.1 | 3 - 4 | 743 | 80 | |
NM_001206916.1 | NP_001193845.1 | 4 - 5 | 517 | 80 | |
NM_001206917.1 | NP_001193846.1 | 4 - 5 | 484 | 80 | |
XM_011538727.2 | XP_011537029.1 | 5 - 6 | 602 | 80 | |
XM_011538728.2 | XP_011537030.1 | 4 - 5 | 1233 | 80 | |
XM_017019957.1 | XP_016875446.1 | 5 - 6 | 1159 | 80 | |
XM_017019958.1 | XP_016875447.1 | 5 - 6 | 1364 | 80 | |
XM_017019959.1 | XP_016875448.1 | 3 - 4 | 362 | 80 | |
GenBank mRNA | AK122911.1 | - | 5 - 6 | 1530 | 80 |
AK289709.1 | - | 4 - 5 | 533 | 80 | |
AK297639.1 | - | 4 - 5 | 504 | 80 | |
AK300901.1 | - | 4 - 5 | 436 | 80 | |
AK304537.1 | - | 4 - 5 | 465 | 80 | |
AK316323.1 | - | 4 - 5 | 500 | 80 | |
AL534844.3 | - | 4 - 5 | 859 | 80 | |
AL702978.1 | - | 4 - 5 | 485 | 80 | |
BC041811.1 | - | 4 - 5 | 606 | 80 | |
CD013906.1 | - | 3 - 4 | 305 | 80 | |
U07139.1 | - | 4 - 5 | 441 | 80 | |
X76555.1 | - | 4 - 5 | 445 | 80 | |
X76556.1 | - | 4 - 5 | 434 | 80 |