Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022072.3 | NP_071355.1 | 4 - 5 | 735 | 118 |
GenBank mRNA | AF169972.1 | - | 4 - 5 | 744 | 118 |
AK026262.1 | - | 4 - 5 | 727 | 118 | |
BC020602.1 | - | 4 - 5 | 735 | 118 | |
BC057238.1 | - | 4 - 5 | 739 | 118 |