Assay Design
Gene Transcripts
- Gene Symbol
- MUC2
- Entrez Gene ID
-
4583
- Gene Name
- mucin 2, oligomeric mucus/gel-forming
- Gene Aliases
-
MLP, MUC-2, SMUC
- Chromosome Location
-
Chr.11: 1074875 - 1110508 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_002457.3 |
NP_002448.3 |
- |
578 |
76 |
GenBank mRNA |
L21998.1 |
- |
- |
578 |
76 |
View Full Product Details