Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003335.2 | NP_003326.2 | 12 - 13 | 1632 | 100 |
XM_005265430.2 | XP_005265487.1 | 12 - 13 | 1616 | 100 | |
XM_011534072.1 | XP_011532374.1 | 11 - 12 | 1525 | 100 | |
GenBank mRNA | BC006378.2 | - | 12 - 13 | 1557 | 100 |
BT007026.1 | - | 12 - 13 | 1461 | 100 | |
L13852.1 | - | 12 - 13 | 1622 | 100 |