Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013351.1 | NP_037483.1 | 5 - 6 | 1193 | 94 |
XM_011524698.2 | XP_011523000.1 | 6 - 7 | 1260 | 94 | |
GenBank mRNA | AF093098.1 | - | 5 - 6 | 1193 | 94 |
AF241243.2 | - | 5 - 6 | 1028 | 94 | |
AK223634.1 | - | 5 - 6 | 1186 | 94 | |
BC039739.2 | - | 5 - 6 | 1158 | 94 |