Assay Design



Gene Transcripts

Gene Symbol
LRRC59
Entrez Gene ID
55379
Gene Name
leucine rich repeat containing 59
Gene Aliases
PRO1855, p34
Chromosome Location
Chr.17: 50381233 - 50397553 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018509.3 NP_060979.2 4 - 5 671 73
GenBank mRNA AK025328.1 - 4 - 5 671 73
AK316593.1 - 2 - 3 173 73
BC017168.1 - 4 - 5 629 73
BC052279.1 - 4 - 5 589 73

View Full Product Details