Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173689.6 | NP_775960.4 | 8 - 9 | 2528 | 79 |
NR_104603.1 | 3 - 4 | 1548 | 79 | ||
XM_005251934.2 | XP_005251991.1 | 3 - 4 | 1548 | 79 | |
XM_011518556.2 | XP_011516858.1 | 8 - 9 | 2528 | 79 | |
XM_011518557.2 | XP_011516859.1 | 8 - 9 | 2530 | 79 | |
XM_011518558.2 | XP_011516860.1 | 9 - 10 | 2890 | 79 | |
GenBank mRNA | AK123000.1 | - | 8 - 9 | 2518 | 79 |
AK126775.1 | - | 3 - 4 | 1548 | 79 | |
AY720432.1 | - | 8 - 9 | 2434 | 79 |