Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004944.3 | NP_004935.1 | 2 - 3 | 328 | 141 |
GenBank mRNA | AF047354.1 | - | 2 - 3 | 301 | 141 |
AK131323.1 | - | 5 - 6 | 823 | 141 | |
AK313303.1 | - | 2 - 3 | 294 | 141 | |
BC015831.1 | - | 2 - 3 | 298 | 141 | |
BI521582.1 | - | 2 - 3 | 538 | 141 | |
U56814.1 | - | 2 - 3 | 255 | 141 |