Assay Design



Gene Transcripts

Gene Symbol
SLC37A4
Entrez Gene ID
2542
Gene Name
solute carrier family 37 member 4
Gene Aliases
G6PT1, G6PT2, G6PT3, GSD1b, GSD1c, GSD1d, PRO0685, TRG-19, TRG19
Chromosome Location
Chr.11: 119024351 - 119030906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164279.1 NP_001157751.1 3 - 4 329 132
GenBank mRNA AK294993.1 - 3 - 4 465 132
AK300646.1 - 3 - 4 295 132

View Full Product Details