Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012112.4 | NP_036244.2 | 15 - 16 | 2527 | 92 |
XM_011528697.2 | XP_011526999.1 | 15 - 16 | 2085 | 92 | |
XM_011528699.2 | XP_011527001.1 | 14 - 15 | 2273 | 92 | |
XM_011528700.2 | XP_011527002.1 | 14 - 15 | 2006 | 92 | |
GenBank mRNA | AB024704.1 | - | 15 - 16 | 2300 | 92 |
AB027467.1 | - | 14 - 15 | 1936 | 92 | |
AF098158.1 | - | 15 - 16 | 2320 | 92 | |
AF146731.1 | - | 14 - 15 | 1933 | 92 | |
AF244547.1 | - | 12 - 13 | 1707 | 92 | |
AF287265.1 | - | 16 - 17 | 2401 | 92 | |
AK001391.1 | - | 11 - 12 | 1610 | 92 | |
AK093379.1 | - | 4 - 5 | 1001 | 92 | |
AK312549.1 | - | 15 - 16 | 2305 | 92 | |
AL117534.1 | - | 13 - 14 | 1771 | 92 | |
AY734680.1 | - | 13 - 14 | 1829 | 92 | |
BC004136.1 | - | 15 - 16 | 2257 | 92 | |
BC011357.1 | - | 2 - 3 | 229 | 92 | |
BC020207.1 | - | 15 - 16 | 2286 | 92 |