Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012112.4 | NP_036244.2 | 2 - 3 | 621 | 98 |
XM_011528697.2 | XP_011526999.1 | 2 - 3 | 179 | 98 | |
XM_011528700.2 | XP_011527002.1 | 1 - 2 | 100 | 98 | |
GenBank mRNA | AB024704.1 | - | 2 - 3 | 394 | 98 |
AB027467.1 | - | 1 - 2 | 30 | 98 | |
AF098158.1 | - | 2 - 3 | 414 | 98 | |
AF287265.1 | - | 2 - 3 | 387 | 98 | |
AK312549.1 | - | 2 - 3 | 399 | 98 | |
BC004136.1 | - | 2 - 3 | 351 | 98 | |
BC020207.1 | - | 2 - 3 | 380 | 98 | |
BU181208.1 | - | 2 - 3 | 463 | 98 | |
CB960160.1 | - | - | 477 | 98 |