Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012112.4 | NP_036244.2 | 7 - 8 | 1305 | 74 |
XM_011528697.2 | XP_011526999.1 | 7 - 8 | 863 | 74 | |
XM_011528699.2 | XP_011527001.1 | 6 - 7 | 1051 | 74 | |
XM_011528700.2 | XP_011527002.1 | 6 - 7 | 784 | 74 | |
GenBank mRNA | AB024704.1 | - | 7 - 8 | 1078 | 74 |
AB027467.1 | - | 6 - 7 | 714 | 74 | |
AF098158.1 | - | 7 - 8 | 1098 | 74 | |
AF146731.1 | - | 6 - 7 | 711 | 74 | |
AF244547.1 | - | 4 - 5 | 485 | 74 | |
AF287265.1 | - | 7 - 8 | 1071 | 74 | |
AK001391.1 | - | 3 - 4 | 388 | 74 | |
AK312549.1 | - | 7 - 8 | 1083 | 74 | |
AL117534.1 | - | 5 - 6 | 549 | 74 | |
AY734680.1 | - | 5 - 6 | 607 | 74 | |
BC004136.1 | - | 7 - 8 | 1035 | 74 | |
BC020207.1 | - | 7 - 8 | 1064 | 74 |