Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012112.4 | NP_036244.2 | 9 - 10 | 1586 | 140 |
XM_011528697.2 | XP_011526999.1 | 9 - 10 | 1144 | 140 | |
XM_011528699.2 | XP_011527001.1 | 8 - 9 | 1332 | 140 | |
XM_011528700.2 | XP_011527002.1 | 8 - 9 | 1065 | 140 | |
GenBank mRNA | AB024704.1 | - | 9 - 10 | 1359 | 140 |
AB027467.1 | - | 8 - 9 | 995 | 140 | |
AF098158.1 | - | 9 - 10 | 1379 | 140 | |
AF146731.1 | - | 8 - 9 | 992 | 140 | |
AF244547.1 | - | 6 - 7 | 766 | 140 | |
AF287265.1 | - | 9 - 10 | 1352 | 140 | |
AK001391.1 | - | 5 - 6 | 669 | 140 | |
AK312549.1 | - | 9 - 10 | 1364 | 140 | |
AL117534.1 | - | 7 - 8 | 830 | 140 | |
AY734680.1 | - | 7 - 8 | 888 | 140 | |
BC004136.1 | - | 9 - 10 | 1316 | 140 | |
BC020207.1 | - | 9 - 10 | 1345 | 140 |