Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003119.3 | NP_003110.1 | 16 - 17 | 2206 | 91 |
GenBank mRNA | AF034795.1 | - | 11 - 12 | 1518 | 91 |
AK094627.1 | - | 4 - 5 | 1160 | 91 | |
BC035929.2 | - | 15 - 16 | 1989 | 91 | |
BC036104.1 | - | 16 - 17 | 2191 | 91 | |
Y16610.1 | - | 16 - 17 | 2188 | 91 |