Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042771.2 | NP_001036236.1 | 4 - 5 | 405 | 53 |
NM_005356.4 | NP_005347.3 | 4 - 5 | 422 | 53 | |
XM_011541453.2 | XP_011539755.1 | 3 - 4 | 596 | 53 | |
XM_011541454.2 | XP_011539756.1 | 3 - 4 | 577 | 53 | |
XM_011541455.2 | XP_011539757.1 | 4 - 5 | 422 | 53 | |
GenBank mRNA | AF228313.1 | - | 3 - 4 | 245 | 53 |
AJ865079.1 | - | 2 - 3 | 458 | 53 | |
AK098027.1 | - | 3 - 4 | 272 | 53 | |
AK303073.1 | - | 3 - 4 | 475 | 53 | |
BC013200.1 | - | 4 - 5 | 384 | 53 | |
DA007906.1 | - | 4 - 5 | 422 | 53 | |
DB122062.1 | - | 4 - 5 | 406 | 53 | |
DB129997.1 | - | 4 - 5 | 343 | 53 | |
KU178161.1 | - | 3 - 4 | 284 | 53 | |
KU178162.1 | - | 3 - 4 | 284 | 53 | |
KU178163.1 | - | 3 - 4 | 284 | 53 | |
U07236.1 | - | 4 - 5 | 310 | 53 | |
X05027.1 | - | 4 - 5 | 376 | 53 |