Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031955.5 | NP_114161.3 | 1 - 2 | 172 | 114 |
XM_006713778.2 | XP_006713841.1 | 1 - 2 | 146 | 114 | |
XM_017007308.1 | XP_016862797.1 | 1 - 2 | 146 | 114 | |
GenBank mRNA | AF345909.2 | - | 1 - 2 | 118 | 113 |
AF404758.1 | - | 1 - 2 | 118 | 113 | |
AK093041.1 | - | 1 - 2 | 120 | 114 | |
BI829426.1 | - | 1 - 2 | 172 | 114 | |
DB040560.1 | - | 1 - 2 | 146 | 114 |