Assay Design



Gene Transcripts

Gene Symbol
SPATA16
Entrez Gene ID
83893
Gene Name
spermatogenesis associated 16
Gene Aliases
NYD-SP12, SPGF6
Chromosome Location
Chr.3: 172889357 - 173141268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031955.5 NP_114161.3 4 - 5 1032 83
XM_006713778.2 XP_006713841.1 4 - 5 1006 83
XM_017007308.1 XP_016862797.1 4 - 5 1006 83
GenBank mRNA AK093041.1 - 4 - 5 980 83
BC034496.2 - 4 - 5 934 83

View Full Product Details