Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005791.2 | NP_005782.1 | 4 - 5 | 1440 | 161 |
GenBank mRNA | AK056862.1 | - | 4 - 5 | 1440 | 161 |
AK310931.1 | - | 4 - 5 | 1122 | 161 | |
BC041084.1 | - | 4 - 5 | 1090 | 161 | |
BC126389.1 | - | 4 - 5 | 1377 | 161 | |
X98494.1 | - | 4 - 5 | 1063 | 161 |