Assay Design



Gene Transcripts

Gene Symbol
MPHOSPH10
Entrez Gene ID
10199
Gene Name
M-phase phosphoprotein 10
Gene Aliases
CT90, MPP10, MPP10P, PPP1R106
Chromosome Location
Chr.2: 71130314 - 71150102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005791.2 NP_005782.1 5 - 6 1592 105
GenBank mRNA AK056862.1 - 5 - 6 1592 105
BC041084.1 - 5 - 6 1242 105
BC126389.1 - 5 - 6 1529 105
X98494.1 - 5 - 6 1215 105

View Full Product Details