Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005791.2 | NP_005782.1 | 7 - 8 | 1791 | 153 |
GenBank mRNA | AK056862.1 | - | 7 - 8 | 1791 | 153 |
BC041084.1 | - | 7 - 8 | 1441 | 153 | |
BC126389.1 | - | 7 - 8 | 1728 | 153 | |
X98494.1 | - | 7 - 8 | 1414 | 153 |