Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077488.3 | NP_001070956.1 | 3 - 5 | 675 | 101 |
NM_016592.3 | NP_057676.1 | 3 - 5 | 1281 | 101 | |
XM_017027812.1 | XP_016883301.1 | 3 - 5 | 2732 | 101 | |
GenBank mRNA | AF105253.1 | - | 3 - 5 | 1259 | 101 |
BC008855.2 | - | 4 - 6 | 393 | 101 | |
M14631.1 | - | 3 - 5 | 218 | 101 |