Assay Design



Gene Transcripts

Gene Symbol
PIWIL4
Entrez Gene ID
143689
Gene Name
piwi like RNA-mediated gene silencing 4
Gene Aliases
HIWI2, MIWI2
Chromosome Location
Chr.11: 94567308 - 94621421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152431.2 NP_689644.2 7 - 8 1124 91
GenBank mRNA AB079366.1 - 7 - 8 1084 91
AK093475.1 - 7 - 8 1064 91
AK096837.1 - 3 - 4 382 91
AK293616.1 - 7 - 8 976 91
BC031060.1 - 7 - 8 916 91
CR749642.1 - 7 - 8 1052 91

View Full Product Details