Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144925.2 | NP_001138397.1 | 3 - 4 | 676 | 106 |
XM_011529568.2 | XP_011527870.1 | 5 - 6 | 685 | 106 | |
XM_017028349.1 | XP_016883838.1 | 3 - 4 | 676 | 106 | |
GenBank mRNA | AK096355.1 | - | 3 - 4 | 719 | 106 |