Assay Design



Gene Transcripts

Gene Symbol
DEF6
Entrez Gene ID
50619
Gene Name
DEF6, guanine nucleotide exchange factor
Gene Aliases
IBP, SLAT, SWAP70L
Chromosome Location
Chr.6: 35297818 - 35321771 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022047.3 NP_071330.3 4 - 5 696 60
GenBank mRNA AJ276095.1 - 4 - 5 657 60
AK314756.1 - 4 - 5 671 60
AY241694.1 - 4 - 5 696 60
BC017504.1 - 4 - 5 662 60

View Full Product Details