Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003747.2 | NP_003738.2 | 15 - 16 | 2319 | 117 |
XM_006716263.3 | XP_006716326.1 | 15 - 16 | 3025 | 117 | |
XM_011543845.2 | XP_011542147.1 | 15 - 16 | 3025 | 117 | |
XM_011543846.2 | XP_011542148.1 | 15 - 16 | 3025 | 117 | |
GenBank mRNA | AB209339.1 | - | 14 - 15 | 1499 | 117 |
AF082556.1 | - | 15 - 16 | 2319 | 117 | |
AF082557.1 | - | 14 - 15 | 1615 | 117 | |
AK302534.1 | - | 15 - 16 | 1993 | 117 |