Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003747.2 | NP_003738.2 | 7 - 8 | 1274 | 116 |
XM_006716263.3 | XP_006716326.1 | 7 - 8 | 1980 | 116 | |
XM_011543845.2 | XP_011542147.1 | 7 - 8 | 1980 | 116 | |
XM_011543846.2 | XP_011542148.1 | 7 - 8 | 1980 | 116 | |
XM_011543847.2 | XP_011542149.1 | 7 - 8 | 1979 | 116 | |
GenBank mRNA | AB209339.1 | - | 6 - 7 | 454 | 116 |
AF082556.1 | - | 7 - 8 | 1274 | 116 | |
AF082557.1 | - | 6 - 7 | 570 | 116 | |
AK301798.1 | - | 7 - 8 | 1195 | 116 | |
AK302031.1 | - | 7 - 8 | 1295 | 116 | |
AK302534.1 | - | 7 - 8 | 948 | 116 | |
AL706619.1 | - | 5 - 6 | 358 | 116 | |
BC098394.1 | - | 7 - 8 | 1297 | 116 |