Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000316.2 | NP_000307.1 | 10 - 11 | 1189 | 89 |
NM_001184744.1 | NP_001171673.1 | 8 - 9 | 1052 | 89 | |
XM_005265344.3 | XP_005265401.1 | 8 - 9 | 2986 | 89 | |
XM_011533967.2 | XP_011532269.1 | 10 - 11 | 1284 | 89 | |
XM_011533968.2 | XP_011532270.1 | 9 - 10 | 1131 | 89 | |
XM_017006932.1 | XP_016862421.1 | 9 - 10 | 1236 | 89 | |
XM_017006933.1 | XP_016862422.1 | 9 - 10 | 1338 | 89 | |
XM_017006934.1 | XP_016862423.1 | 10 - 11 | 1254 | 89 | |
GenBank mRNA | AY449732.1 | - | 8 - 9 | 986 | 89 |
BC110388.1 | - | 8 - 9 | 1052 | 89 | |
BC112221.1 | - | 10 - 11 | 1189 | 89 | |
BC112247.1 | - | 10 - 11 | 1189 | 89 | |
L04308.1 | - | 8 - 9 | 1014 | 89 | |
U17418.1 | - | 10 - 11 | 1204 | 89 | |
X68596.1 | - | 10 - 11 | 1160 | 89 |