Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000316.2 | NP_000307.1 | 2 - 3 | 155 | 84 |
XM_011533967.2 | XP_011532269.1 | 2 - 3 | 211 | 84 | |
XM_017006934.1 | XP_016862423.1 | 2 - 3 | 181 | 84 | |
GenBank mRNA | BC112221.1 | - | 2 - 3 | 155 | 84 |
BC112247.1 | - | 2 - 3 | 155 | 84 | |
U17418.1 | - | 2 - 3 | 170 | 84 | |
X68596.1 | - | 2 - 3 | 126 | 84 |