Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000593.5 | NP_000584.2 | 2 - 3 | 1051 | 134 |
NM_001292022.1 | NP_001278951.1 | 2 - 3 | 311 | 134 | |
GenBank mRNA | AB012644.1 | - | 2 - 3 | 715 | 134 |
AK302340.1 | - | 2 - 3 | 282 | 134 | |
AK313762.1 | - | 2 - 3 | 1087 | 134 | |
AY523970.1 | - | 2 - 3 | 922 | 134 | |
AY523971.2 | - | 2 - 3 | 922 | 134 | |
BC014081.2 | - | 2 - 3 | 1051 | 134 | |
BX648013.1 | - | 2 - 3 | 1059 | 134 | |
DC354917.1 | - | 2 - 3 | 311 | 134 | |
KU178346.1 | - | 2 - 3 | 896 | 134 | |
L21204.1 | - | 2 - 3 | 716 | 134 | |
L21205.1 | - | 2 - 3 | 716 | 134 | |
L21206.1 | - | 2 - 3 | 716 | 134 | |
L21207.1 | - | 2 - 3 | 716 | 134 | |
L21208.1 | - | 2 - 3 | 716 | 134 | |
X57521.1 | - | 2 - 3 | 350 | 134 | |
X57522.1 | - | 2 - 3 | 926 | 134 |