Assay Design



Gene Transcripts

Gene Symbol
PTK7
Entrez Gene ID
5754
Gene Name
protein tyrosine kinase 7 (inactive)
Gene Aliases
CCK-4, CCK4
Chromosome Location
Chr.6: 43076268 - 43161720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002821.4 NP_002812.2 1 - 1 228 87
NM_152880.3 NP_690619.1 1 - 1 228 87
NM_152881.3 NP_690620.1 1 - 1 228 87
NM_152882.3 NP_690621.1 1 - 1 228 87
NR_072997.1 1 - 1 228 87
NR_072998.1 1 - 1 228 87
GenBank mRNA AB209350.1 - 1 - 1 127 87
AF531868.1 - 1 - 1 154 87
AF531869.1 - 1 - 1 154 87
AF531870.1 - 1 - 1 154 87
AF531871.1 - 1 - 1 154 87
AF531872.1 - 1 - 1 154 87
AK291016.1 - 1 - 1 60 87
AK292589.1 - 1 - 1 155 87
BC046109.2 - 1 - 1 168 87
BC071557.1 - 1 - 1 182 87
BM811107.1 - - 205 87
CA389442.1 - 1 - 1 228 87
U40271.2 - 1 - 1 154 87

View Full Product Details