Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270398.1 | NP_001257327.1 | 10 - 11 | 1714 | 71 |
NM_002821.4 | NP_002812.2 | 10 - 11 | 1841 | 71 | |
NM_152882.3 | NP_690621.1 | 10 - 11 | 1841 | 71 | |
NR_072997.1 | 9 - 10 | 1650 | 71 | ||
NR_072998.1 | 10 - 11 | 1841 | 71 | ||
GenBank mRNA | AB209350.1 | - | 10 - 11 | 1740 | 71 |
AF531868.1 | - | 10 - 11 | 1767 | 71 | |
AF531871.1 | - | 10 - 11 | 1767 | 71 | |
AF531872.1 | - | 10 - 11 | 1767 | 71 | |
AK292589.1 | - | 10 - 11 | 1768 | 71 | |
AK296931.1 | - | 11 - 12 | 1632 | 71 | |
AK296953.1 | - | 10 - 11 | 1649 | 71 | |
BC071557.1 | - | 10 - 11 | 1795 | 71 | |
DA048438.1 | - | 2 - 3 | 182 | 71 | |
U33635.1 | - | 10 - 11 | 1812 | 71 | |
U40271.2 | - | 10 - 11 | 1767 | 71 |