Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270398.1 | NP_001257327.1 | 14 - 15 | 2343 | 77 |
NM_002821.4 | NP_002812.2 | 14 - 15 | 2470 | 77 | |
NM_152880.3 | NP_690619.1 | 13 - 14 | 2350 | 77 | |
NM_152881.3 | NP_690620.1 | 11 - 12 | 2080 | 77 | |
NM_152882.3 | NP_690621.1 | 13 - 14 | 2302 | 77 | |
NR_072997.1 | 13 - 14 | 2274 | 77 | ||
NR_072998.1 | 14 - 15 | 2470 | 77 | ||
XM_011514765.2 | XP_011513067.1 | 13 - 14 | 2218 | 77 | |
XM_011514766.2 | XP_011513068.1 | 11 - 12 | 1948 | 77 | |
GenBank mRNA | AB209350.1 | - | 13 - 14 | 3486 | 77 |
AF531868.1 | - | 14 - 15 | 2396 | 77 | |
AF531869.1 | - | 13 - 14 | 2276 | 77 | |
AF531870.1 | - | 11 - 12 | 2006 | 77 | |
AF531871.1 | - | 13 - 14 | 2228 | 77 | |
AF531872.1 | - | 14 - 15 | 2396 | 77 | |
AK055648.1 | - | 1 - 2 | 596 | 77 | |
AK131487.1 | - | 1 - 2 | 1271 | 77 | |
AK291016.1 | - | 13 - 14 | 2182 | 77 | |
AK292589.1 | - | 14 - 15 | 2397 | 77 | |
BC071557.1 | - | 14 - 15 | 2424 | 77 | |
BQ881986.1 | - | 2 - 3 | 271 | 77 | |
U33635.1 | - | 14 - 15 | 2441 | 77 | |
U40271.2 | - | 14 - 15 | 2396 | 77 |