Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099436.3 | NP_001092906.3 | 8 - 9 | 1063 | 59 |
NM_001284364.2 | NP_001271293.2 | 8 - 9 | 1063 | 59 | |
NM_001284365.2 | NP_001271294.1 | 8 - 9 | 986 | 59 | |
NR_104307.2 | 7 - 8 | 1244 | 59 | ||
NR_104308.2 | 7 - 8 | 1244 | 59 | ||
NR_104309.2 | 7 - 8 | 1244 | 59 | ||
XM_005254289.2 | XP_005254346.1 | 9 - 10 | 1078 | 59 | |
XM_017022068.1 | XP_016877557.1 | 9 - 10 | 1112 | 59 | |
XM_017022069.1 | XP_016877558.1 | 9 - 10 | 1116 | 59 | |
XM_017022070.1 | XP_016877559.1 | 9 - 10 | 1129 | 59 | |
XM_017022071.1 | XP_016877560.1 | 9 - 10 | 1116 | 59 | |
XM_017022072.1 | XP_016877561.1 | 8 - 9 | 1292 | 59 | |
GenBank mRNA | AK075047.1 | - | 7 - 8 | 1153 | 59 |
AK293181.1 | - | 8 - 9 | 916 | 59 | |
AK294241.1 | - | 8 - 9 | 997 | 59 | |
AK294245.1 | - | 8 - 9 | 993 | 59 | |
AK294251.1 | - | 8 - 9 | 997 | 59 | |
AK298879.1 | - | 8 - 9 | 920 | 59 | |
AK299380.1 | - | 8 - 9 | 990 | 59 | |
AL117482.1 | - | 7 - 8 | 1379 | 59 | |
AL360256.1 | - | 3 - 4 | 656 | 59 | |
BC036117.1 | - | 7 - 8 | 1215 | 59 | |
BC048289.1 | - | 7 - 8 | 1172 | 59 | |
BC056423.1 | - | 7 - 8 | 1162 | 59 | |
BC157884.1 | - | 8 - 9 | 1045 | 59 |