Assay Design
Gene Transcripts
- Gene Symbol
- GLS
- Entrez Gene ID
-
2744
- Gene Name
- glutaminase
- Gene Aliases
-
AAD20, GAC, GAM, GLS1, KGA
- Chromosome Location
-
Chr.2: 190880821 - 190965552 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK092974.1 |
- |
1 - 1 |
793 |
106 |
View Full Product Details