Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001326559.1 | NP_001313488.1 | 17 - 18 | 3116 | 98 |
NM_005070.3 | NP_005061.2 | 17 - 18 | 3035 | 98 | |
NM_201574.2 | NP_963868.2 | 17 - 18 | 3056 | 98 | |
NR_048551.1 | 17 - 18 | 3060 | 98 | ||
XM_005246789.4 | XP_005246846.1 | 17 - 18 | 3056 | 98 | |
XM_005246790.4 | XP_005246847.1 | 12 - 13 | 2327 | 98 | |
XM_011511665.2 | XP_011509967.1 | 17 - 18 | 3247 | 98 | |
XM_011511667.2 | XP_011509969.1 | 8 - 9 | 1409 | 98 | |
GenBank mRNA | AK226122.1 | - | 16 - 17 | 2635 | 98 |
AK289974.1 | - | 17 - 18 | 2969 | 98 | |
AK299109.1 | - | 16 - 17 | 2498 | 98 | |
AK299595.1 | - | 9 - 10 | 1853 | 98 | |
AK314083.1 | - | 17 - 18 | 3004 | 98 | |
AY142112.1 | - | 17 - 18 | 3066 | 98 | |
BC136384.1 | - | 16 - 17 | 2802 | 98 | |
BC146656.1 | - | 17 - 18 | 2903 | 98 | |
BC171760.1 | - | 16 - 17 | 2802 | 98 | |
L27213.1 | - | 11 - 12 | 1894 | 98 | |
U05596.1 | - | 16 - 17 | 2777 | 98 |