Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001326559.1 | NP_001313488.1 | 18 - 19 | 3210 | 70 |
NM_005070.3 | NP_005061.2 | 18 - 19 | 3129 | 70 | |
NM_201574.2 | NP_963868.2 | 18 - 19 | 3150 | 70 | |
NR_048551.1 | 18 - 19 | 3154 | 70 | ||
XM_005246789.4 | XP_005246846.1 | 18 - 19 | 3150 | 70 | |
XM_005246790.4 | XP_005246847.1 | 13 - 14 | 2421 | 70 | |
XM_011511665.2 | XP_011509967.1 | 18 - 19 | 3341 | 70 | |
XM_011511667.2 | XP_011509969.1 | 9 - 10 | 1503 | 70 | |
GenBank mRNA | AK226122.1 | - | 17 - 18 | 2729 | 70 |
AK299109.1 | - | 17 - 18 | 2592 | 70 | |
AK299595.1 | - | 10 - 11 | 1947 | 70 | |
AK314083.1 | - | 18 - 19 | 3098 | 70 | |
AY142112.1 | - | 18 - 19 | 3160 | 70 | |
BC136384.1 | - | 17 - 18 | 2896 | 70 | |
BC146656.1 | - | 18 - 19 | 2997 | 70 | |
BC171760.1 | - | 17 - 18 | 2896 | 70 | |
U05596.1 | - | 17 - 18 | 2871 | 70 |