Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001326559.1 | NP_001313488.1 | 21 - 22 | 3796 | 67 |
NM_005070.3 | NP_005061.2 | 21 - 22 | 3715 | 67 | |
NM_201574.2 | NP_963868.2 | 21 - 22 | 3736 | 67 | |
NR_048551.1 | 21 - 22 | 3740 | 67 | ||
XM_005246789.4 | XP_005246846.1 | 21 - 22 | 3736 | 67 | |
XM_005246790.4 | XP_005246847.1 | 16 - 17 | 3007 | 67 | |
XM_011511665.2 | XP_011509967.1 | 21 - 22 | 3927 | 67 | |
XM_011511667.2 | XP_011509969.1 | 12 - 13 | 2089 | 67 | |
GenBank mRNA | AK226122.1 | - | 20 - 21 | 3315 | 67 |
AK289974.1 | - | 21 - 22 | 3649 | 67 | |
AK299109.1 | - | 20 - 21 | 3178 | 67 | |
AK299595.1 | - | 13 - 14 | 2533 | 67 | |
AK314083.1 | - | 21 - 22 | 3684 | 67 | |
AY142112.1 | - | 21 - 22 | 3746 | 67 | |
BC136384.1 | - | 20 - 21 | 3482 | 67 | |
BC146656.1 | - | 21 - 22 | 3583 | 67 | |
BC171760.1 | - | 20 - 21 | 3482 | 67 | |
L27213.1 | - | 15 - 16 | 2574 | 67 | |
U05596.1 | - | 20 - 21 | 3457 | 67 |