Assay Design



Gene Transcripts

Gene Symbol
NEU4
Entrez Gene ID
129807
Gene Name
neuraminidase 4
Gene Aliases
-
Chromosome Location
Chr.2: 241809065 - 241817413 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167602.2 NP_001161074.1 1 - 2 527 110
GenBank mRNA AK091038.1 - 1 - 2 524 110

View Full Product Details