Assay Design
Gene Transcripts
- Gene Symbol
- LOC100288721
- Entrez Gene ID
-
100288721
- Gene Name
- uncharacterized LOC100288721
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC015772.1 |
- |
1 - 1 |
455 |
140 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG764444.1 |
- |
1 |
455 |
|
Chr. 3 - 109176604 - 109177366 [-] on Build GRCh38 |
140 |
GenBankmRNA |
BG768338.1 |
- |
1 |
455 |
|
Chr. 3 - 109176592 - 109177366 [-] on Build GRCh38 |
140 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC401081 |
FLJ22763 |
|
-26.5kb
|
Anti-sense |
Chr. - 109136714 - 109150109 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 488 |
LINC00488 |
C3orf66 |
799.0
|
Anti-sense |
Chr. - 109178165 - 109185261 on Build GRCh38 |
View Full Product Details