Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159969.1 | NP_001153441.1 | 2 - 3 | 496 | 132 |
NM_001981.2 | NP_001972.1 | 14 - 15 | 1365 | 132 | |
XM_005270618.2 | XP_005270675.1 | 13 - 14 | 1306 | 132 | |
XM_017000615.1 | XP_016856104.1 | 14 - 15 | 1507 | 132 | |
XM_017000616.1 | XP_016856105.1 | 14 - 15 | 1507 | 132 | |
XM_017000617.1 | XP_016856106.1 | 14 - 15 | 1396 | 132 | |
XM_017000618.1 | XP_016856107.1 | 13 - 14 | 1306 | 132 | |
XM_017000619.1 | XP_016856108.1 | 15 - 16 | 1389 | 132 | |
XM_017000620.1 | XP_016856109.1 | 14 - 15 | 1556 | 132 | |
GenBank mRNA | AK294297.1 | - | 3 - 4 | 345 | 132 |
AK313396.1 | - | 14 - 15 | 1314 | 132 | |
BX647676.1 | - | 2 - 3 | 480 | 129 | |
CD657119.1 | - | 1 - 2 | 140 | 132 | |
DQ367924.1 | - | 14 - 15 | 1273 | 132 | |
U07707.1 | - | 14 - 15 | 1294 | 132 | |
Z29064.1 | - | 14 - 15 | 1365 | 132 |