Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001981.2 | NP_001972.1 | 5 - 6 | 404 | 98 |
XM_005270618.2 | XP_005270675.1 | 5 - 6 | 435 | 98 | |
XM_017000615.1 | XP_016856104.1 | 5 - 6 | 435 | 98 | |
XM_017000616.1 | XP_016856105.1 | 5 - 6 | 435 | 98 | |
XM_017000617.1 | XP_016856106.1 | 5 - 6 | 435 | 98 | |
XM_017000618.1 | XP_016856107.1 | 5 - 6 | 435 | 98 | |
XM_017000619.1 | XP_016856108.1 | 5 - 6 | 320 | 98 | |
XM_017000620.1 | XP_016856109.1 | 5 - 6 | 323 | 98 | |
GenBank mRNA | AK129853.1 | - | 5 - 6 | 326 | 98 |
AK313396.1 | - | 5 - 6 | 353 | 98 | |
BC054006.1 | - | 5 - 6 | 322 | 98 | |
DQ367924.1 | - | 5 - 6 | 312 | 98 | |
U07707.1 | - | 5 - 6 | 333 | 98 | |
Z29064.1 | - | 5 - 6 | 404 | 98 |