Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001981.2 | NP_001972.1 | 7 - 8 | 599 | 95 |
XM_005270618.2 | XP_005270675.1 | 7 - 8 | 630 | 95 | |
XM_017000615.1 | XP_016856104.1 | 7 - 8 | 630 | 95 | |
XM_017000616.1 | XP_016856105.1 | 7 - 8 | 630 | 95 | |
XM_017000617.1 | XP_016856106.1 | 7 - 8 | 630 | 95 | |
XM_017000618.1 | XP_016856107.1 | 7 - 8 | 630 | 95 | |
XM_017000619.1 | XP_016856108.1 | 7 - 8 | 515 | 95 | |
XM_017000620.1 | XP_016856109.1 | 7 - 8 | 518 | 95 | |
GenBank mRNA | AK129853.1 | - | 7 - 8 | 521 | 95 |
AK313396.1 | - | 7 - 8 | 548 | 95 | |
BC054006.1 | - | 7 - 8 | 517 | 95 | |
DQ367924.1 | - | 7 - 8 | 507 | 95 | |
U07707.1 | - | 7 - 8 | 528 | 95 | |
Z29064.1 | - | 7 - 8 | 599 | 95 |