Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190481.1 | NP_001177410.1 | 20 - 21 | 3446 | 94 |
NM_022111.3 | NP_071394.2 | 21 - 22 | 3638 | 94 | |
XM_011541938.2 | XP_011540240.1 | 21 - 22 | 3661 | 94 | |
XM_011541939.2 | XP_011540241.1 | 21 - 22 | 3655 | 94 | |
XM_011541940.2 | XP_011540242.1 | 21 - 22 | 3655 | 94 | |
XM_011541941.2 | XP_011540243.1 | 20 - 21 | 3493 | 94 | |
GenBank mRNA | AF297866.1 | - | 21 - 22 | 3638 | 94 |
BC113116.1 | - | 21 - 22 | 3631 | 94 | |
BC115025.1 | - | 20 - 21 | 3439 | 94 | |
BC115026.1 | - | 21 - 22 | 3631 | 94 | |
BC137279.1 | - | 21 - 22 | 3617 | 94 | |
BC140789.1 | - | 21 - 22 | 3617 | 94 |