Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166588.2 | NP_001160060.1 | 9 - 10 | 1292 | 57 |
NM_001261418.1 | NP_001248347.1 | 9 - 10 | 1292 | 57 | |
NM_020365.4 | NP_065098.1 | 9 - 10 | 1292 | 57 | |
XM_017002745.1 | XP_016858234.1 | 9 - 10 | 1180 | 57 | |
XM_017002746.1 | XP_016858235.1 | 10 - 11 | 1259 | 57 | |
XM_017002747.1 | XP_016858236.1 | 10 - 11 | 1253 | 57 | |
GenBank mRNA | AF257077.1 | - | 9 - 10 | 1155 | 57 |
AK022399.1 | - | 9 - 10 | 1154 | 57 | |
AK314668.1 | - | 9 - 10 | 1165 | 57 | |
AL834288.1 | - | 9 - 10 | 1157 | 57 | |
AW972575.1 | - | 1 - 2 | 57 | ||
BC018728.1 | - | 9 - 10 | 1150 | 57 | |
BC021017.1 | - | 9 - 10 | 1143 | 57 | |
BQ776018.1 | - | 3 - 4 | 57 | ||
CR457285.1 | - | 5 - 6 | 546 | 57 | |
CX781577.1 | - | 1 - 2 | 57 |